Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other METTL23 SIRNA Assays ›
Gene Symbol:
METTL23
Gene Name:
methyltransferase like 23
Gene Aliases:
C17orf95, MRT44
Chromosome Location:
Chr.17: 76725874 - 76733881 on Build GRCh38
UniGene ID:
Hs.74655
Species:
Human
siRNA ID
HSS174720
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: METTL23
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080510.4
NP_001073979.3
4
752
NM_001206983.2
NP_001193912.1
4
787
NM_001206984.2
NP_001193913.1
4
453
NM_001206985.2
NP_001193914.1
3
647
NM_001206986.2
NP_001193915.1
3
682
NM_001206987.2
NP_001193916.1
3
826
NM_001302703.1
NP_001289632.1
5
697
NM_001302704.1
NP_001289633.1
4
557
NM_001302705.1
NP_001289634.1
4
436
NR_038193.2
2
409
XM_006721674.3
XP_006721737.1
4
815
XM_006721675.1
XP_006721738.1
4
679
XM_006721676.3
XP_006721739.1
4
714
XM_006721678.3
XP_006721741.1
4
800
XM_006721679.3
XP_006721742.1
3
445
XM_006721680.2
XP_006721743.1
3
354
XM_011524282.1
XP_011522584.1
4
543
XM_017024145.1
XP_016879634.1
5
647
XM_017024146.1
XP_016879635.1
4
557
GenBank mRNA
AL522160.3
-
4
737
BC033998.2
-
3
623
BC045819.1
-
4
714
BC106939.2
-
2
275
BE567652.1
-
4
408
BF575183.1
-
4
454
BI225157.1
-
3
646
BI829268.1
-
3
608
BU176249.1
-
4
439
BX356072.2
-
4
536
BX396749.2
-
5
684
BX403283.2
-
4
563
CN372746.1
-
2
365
HY285382.1
-
2
414
JQ313903.1
-
3
382
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
methylation
methyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0