Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR2C2AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300945.1 | NP_001287874.1 | 4, 5 | 664 | ||
NM_176880.5 | NP_795361.1 | 4, 5 | 664 | |||
GenBank mRNA | AK301426.1 | - | 4, 5 | 592 | ||
AY101377.1 | - | 4, 5 | 508 | |||
BC057837.1 | - | 4, 5 | 643 | 30345563 |