Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PODN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153703.4 | NP_714914.2 | 5 | 832 | ||
XM_005270457.3 | XP_005270514.1 | 6 | 861 | |||
XM_017000294.1 | XP_016855783.1 | 5 | 1182 | |||
GenBank mRNA | AK296649.1 | - | 6 | 727 | ||
AK299375.1 | - | 7 | 917 | |||
AK314096.1 | - | 5 | 426 | |||
AY313607.1 | - | 5 | 671 | |||
AY358294.1 | - | 7 | 948 | |||
BC028292.1 | - | 5 | 655 | 4938380 | ||
BC030608.2 | - | 5 | 673 | 4826932 |