Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TATDN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042552.2 | NP_001036017.1 | 8 | 599 | ||
NM_001042553.2 | NP_001036018.1 | 8 | 599 | |||
NM_001146169.1 | NP_001139641.1 | 8 | 599 | |||
NM_001146170.1 | NP_001139642.1 | 7 | 536 | |||
NM_001146171.1 | NP_001139643.1 | 8 | 599 | |||
XM_017000328.1 | XP_016855817.1 | 7 | 531 | |||
XM_017000329.1 | XP_016855818.1 | 7 | 531 | |||
XM_017000330.1 | XP_016855819.1 | 8 | 594 | |||
GenBank mRNA | AK295335.1 | - | 7 | 524 | ||
AK304557.1 | - | 7 | 494 | |||
AL832248.1 | - | 8 | 541 | |||
BC048115.1 | - | 8 | 510 | |||
BC113638.1 | - | 8 | 543 | 8322690 | ||
BC117485.1 | - | 8 | 543 | 40126036 | ||
BC143956.1 | - | 8 | 543 | 9052474 | ||
BC143957.1 | - | 8 | 543 |