Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL4A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287758.1 | NP_001274687.1 | 20 | 1567 | ||
NM_001287759.1 | NP_001274688.1 | 20 | 1567 | |||
NM_001287760.1 | NP_001274689.1 | 20 | 1567 | |||
NM_001847.3 | NP_001838.2 | 20 | 1462 | |||
NM_033641.3 | NP_378667.1 | 20 | 1567 | |||
XM_006724617.3 | XP_006724680.1 | 20 | 1456 | |||
XM_011530852.2 | XP_011529154.1 | 20 | 1456 | |||
XM_011530853.2 | XP_011529155.1 | 18 | 2172 | |||
XM_011530854.2 | XP_011529156.1 | 20 | 1457 | |||
GenBank mRNA | BC140855.1 | - | 20 | 1463 | 9021723 | |
BC144669.1 | - | 20 | 1463 | 9053200 | ||
CN313332.1 | - | 2 | 105 | |||
CN313341.1 | - | 4 | 285 | |||
D21337.1 | - | 20 | 1567 | |||
L22763.1 | - | 20 | 1445 | |||
U04845.1 | - | 20 | 1445 |