Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: UBR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172070.3 | NP_742067.3 | 37 | 5248 | ||
XM_005246299.4 | XP_005246356.1 | 38 | 5338 | |||
XM_005246300.4 | XP_005246357.1 | 38 | 5281 | |||
XM_006712268.3 | XP_006712331.1 | 38 | 5335 | |||
XM_006712269.3 | XP_006712332.1 | 37 | 5260 | |||
XM_006712270.3 | XP_006712333.2 | 37 | 5194 | |||
XM_006712271.3 | XP_006712334.1 | 37 | 5301 | |||
GenBank mRNA | AB095944.1 | - | 8 | 1358 | ||
AK094741.1 | - | 1 | 20 | |||
AK126998.1 | - | 15 | 1977 | |||
AK127553.1 | - | 11 | 1254 | |||
AK128490.1 | - | 21 | 2728 | |||
AK303062.1 | - | 12 | 1348 | |||
AL834144.1 | - | 9 | 955 | |||
BX649036.1 | - | 13 | 1484 | |||
CR749469.1 | - | 15 | 1750 |