Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102609.2 | NP_001096079.1 | 3 | 250 | ||
NM_001305393.1 | NP_001292322.1 | 3 | 269 | |||
NM_001305394.1 | NP_001292323.1 | 3 | 258 | |||
NM_001305395.1 | NP_001292324.1 | 3 | 250 | |||
NM_001305396.1 | NP_001292325.1 | 3 | 239 | |||
NM_001305397.1 | NP_001292326.1 | 3 | 250 | |||
NR_131091.1 | 3 | 250 | ||||
NR_131092.1 | 2 | 166 | ||||
XM_011534432.2 | XP_011532734.1 | 3 | 721 | |||
XM_017009029.1 | XP_016864518.1 | 3 | 1437 | |||
XM_017009030.1 | XP_016864519.1 | 3 | 1436 | |||
XM_017009031.1 | XP_016864520.1 | 3 | 310 | |||
XM_017009032.1 | XP_016864521.1 | 3 | 234 | |||
XM_017009033.1 | XP_016864522.1 | 3 | 301 | |||
GenBank mRNA | AA758527.1 | - | 2 | 290 | ||
BC030767.1 | - | 3 | 230 | |||
BC092511.1 | - | 2 | 1230 | |||
BU561519.1 | - | 2 | 146 | |||
BU561730.1 | - | 3 | 186 | |||
BX100176.1 | - | 2 | 170 | |||
HY000873.1 | - | 2 | 162 | |||
HY001392.1 | - | 2 | 173 | |||
HY005987.1 | - | 3 | 269 | |||
HY016595.1 | - | 3 | 175 | |||
HY038777.1 | - | 3 | 250 |