Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159587.2 | NP_001153059.1 | 17 | 2079 | ||
NM_001159588.2 | NP_001153060.1 | 16 | 1848 | |||
NM_133493.4 | NP_598000.2 | 17 | 2079 | |||
XM_005248659.3 | XP_005248716.1 | 18 | 2226 | |||
XM_011535473.1 | XP_011533775.1 | 17 | 1924 | |||
XM_017010283.1 | XP_016865772.1 | 15 | 1691 | |||
GenBank mRNA | AF410459.1 | - | 17 | 2060 | ||
AK095888.1 | - | 17 | 2013 | |||
AK123960.1 | - | 3 | 157 | |||
AK313636.1 | - | 17 | 2059 | |||
AL834478.1 | - | 3 | 175 | |||
AY149920.1 | - | 17 | 1948 | |||
AY788891.1 | - | 17 | 1973 | |||
BX641095.1 | - | 16 | 1735 | |||
EF553520.1 | - | 17 | 1978 |