Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178819.3 | NP_848934.1 | 5, 6 | 1556 | ||
XM_011544392.2 | XP_011542694.1 | 6, 7 | 1679 | |||
XM_011544393.1 | XP_011542695.1 | 5, 6 | 1568 | |||
XM_011544394.1 | XP_011542696.1 | 5, 6 | 1605 | |||
XM_017013042.1 | XP_016868531.1 | 6, 7 | 1706 | |||
XM_017013043.1 | XP_016868532.1 | 6, 7 | 1577 | |||
GenBank mRNA | AF317516.1 | - | 4, 5 | 782 | ||
AF406612.1 | - | 5, 6 | 1544 | |||
AL834255.1 | - | 1, 2 | 41 | |||
AY513610.1 | - | 4, 5 | 776 | |||
BC051377.1 | - | 4, 5 | 736 | 6649895 | ||
BC061884.1 | - | 4, 5 | 736 | 6577569 |