Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184767.1 | NP_001171696.1 | 9 | 1581 | ||
NM_153346.4 | NP_699177.2 | 11 | 1854 | |||
XM_011545447.1 | XP_011543749.1 | 11 | 1878 | |||
XM_017029266.1 | XP_016884755.1 | 10 | 1752 | |||
XM_017029267.1 | XP_016884756.1 | 11 | 1878 | |||
GenBank mRNA | AK128155.1 | - | 11 | 1836 | ||
BC037301.2 | - | 11 | 1834 | 4827886 | ||
BC096745.1 | - | 9 | 1514 | 4830576 |