Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRHR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202475.1 | NP_001189404.1 | 6 | 603 | ||
NM_001202481.1 | NP_001189410.1 | 7 | 830 | |||
NM_001202482.1 | NP_001189411.1 | 5 | 694 | |||
NM_001202483.1 | NP_001189412.1 | 5 | 697 | |||
NM_001883.4 | NP_001874.2 | 5 | 697 | |||
XM_011515127.2 | XP_011513429.1 | 5 | 504 | |||
XM_011515128.2 | XP_011513430.1 | 5 | 504 | |||
XM_011515129.2 | XP_011513431.1 | 5 | 504 | |||
XM_017011751.1 | XP_016867240.1 | 5 | 501 | |||
XM_017011752.1 | XP_016867241.1 | 5 | 1641 | |||
GenBank mRNA | AF011406.1 | - | 6 | 603 | ||
AF019381.1 | - | 5 | 561 | |||
AK313661.1 | - | 5 | 658 | |||
AY449734.1 | - | 5 | 452 | |||
BC096830.1 | - | 5 | 461 | 6971903 | ||
EU012439.1 | - | 5 | 452 | |||
EU012440.1 | - | 5 | 452 | |||
EU012441.1 | - | 5 | 452 | |||
EU012442.1 | - | 4 | 363 | |||
EU012443.1 | - | 5 | 449 | |||
FJ200484.1 | - | 4 | 363 | |||
U34587.1 | - | 5 | 452 |
Set Membership: |
G-protein coupled receptor |