Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080621.4 | NP_542188.1 | 2, 3 | 459 | ||
XM_005260199.4 | XP_005260256.1 | 4, 5 | 1596 | |||
XM_006723705.3 | XP_006723768.1 | 3, 4 | 358 | |||
XM_011528565.2 | XP_011526867.1 | 3, 4 | 1228 | |||
XM_017027671.1 | XP_016883160.1 | 3, 4 | 1476 | |||
GenBank mRNA | BC067362.1 | - | 2, 3 | 456 | 6672007 |