Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000496.2 | NP_000487.1 | 5 | 409 | ||
XM_006724141.3 | XP_006724204.1 | 5 | 1568 | |||
XM_011529900.2 | XP_011528202.1 | 6 | 1933 | |||
GenBank mRNA | BC069535.1 | - | 4 | 373 | 7262188 | |
BM667912.1 | - | 2 | 369 | |||
CR456426.1 | - | 4 | 371 | |||
KR052782.1 | - | 5 | 403 | |||
KR052783.1 | - | 5 | 403 | |||
KR052784.1 | - | 5 | 405 | |||
KR052788.1 | - | 4 | 361 | |||
KR052789.1 | - | 4 | 352 | |||
KR052790.1 | - | 4 | 394 | |||
KR052791.1 | - | 3 | 309 | |||
L10035.1 | - | 4 | 373 |
Gene Symbol: CRYBB2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033733.1 | 4 | 501 | |||
NR_033734.1 | 3 | 382 | ||||
GenBank mRNA | BC037884.1 | - | 3 | 369 | ||
BC047380.1 | - | 4 | 504 |