Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MIPOL1 SIRNA Assays ›
Gene Symbol:
MIPOL1
Gene Name:
mirror-image polydactyly 1
Gene Aliases:
CCDC193
Chromosome Location:
Chr.14: 37197888 - 37579207 on Build GRCh38
UniGene ID:
Hs.660396
Species:
Human
siRNA ID
HSS175378
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MIPOL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001195296.1
NP_001182225.1
14
1859
NM_001195297.1
NP_001182226.1
14
1845
NM_138731.6
NP_620059.1
13
1706
XM_011536463.2
XP_011534765.1
15
1984
XM_017020991.1
XP_016876480.1
16
2092
XM_017020992.1
XP_016876481.1
15
1955
XM_017020993.1
XP_016876482.1
15
1965
XM_017020994.1
XP_016876483.1
12
1610
XM_017020995.1
XP_016876484.1
14
1826
XM_017020996.1
XP_016876485.1
14
1842
XM_017020997.1
XP_016876486.1
13
1715
XM_017020998.1
XP_016876487.1
16
2091
XM_017020999.1
XP_016876488.1
17
2167
XM_017021000.1
XP_016876489.1
13
1651
XM_017021001.1
XP_016876490.1
14
1790
XM_017021002.1
XP_016876491.1
10
1241
XM_017021003.1
XP_016876492.1
16
2090
XM_017021004.1
XP_016876493.1
12
1618
XM_017021005.1
XP_016876494.1
13
1713
XM_017021006.1
XP_016876495.1
16
2090
XM_017021007.1
XP_016876496.1
16
2090
XM_017021008.1
XP_016876497.1
13
1649
XM_017021009.1
XP_016876498.1
14
1762
XM_017021010.1
XP_016876499.1
10
1241
XM_017021011.1
XP_016876500.1
11
1466
XM_017021012.1
XP_016876501.1
13
1654
XM_017021013.1
XP_016876502.1
9
1133
XM_017021019.1
XP_016876508.1
6
4556
GenBank mRNA
AK291850.1
-
14
1770
AY059470.1
-
14
1670
BC035870.1
-
7
865
5245356
BC036124.1
-
14
1731
5298587
BC036523.1
-
13
1574
4828885
BX537396.1
-
13
1669
BX647795.1
-
9
1102
KU178867.1
-
6
660
KU178868.1
-
7
708
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-f2nf4:80/10.239.165.207:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0