Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139170.2 | NP_631909.2 | 3 | 664 | ||
XM_005255144.3 | XP_005255201.1 | 2 | 393 | |||
XM_017022975.1 | XP_016878464.1 | 3 | 520 | |||
XM_017022976.1 | XP_016878465.1 | 3 | 282 | |||
GenBank mRNA | AF447587.1 | - | 3 | 638 | ||
AK301794.1 | - | 3 | 490 | |||
BC035024.1 | - | 3 | 614 | 4826923 | ||
BX648126.1 | - | 3 | 450 |