Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001899.2 | NP_001890.1 | 1 | 222 | ||
GenBank mRNA | BC065714.1 | - | 1 | 239 | 30327736 | |
BC074952.2 | - | 1 | 195 | 30915278 | ||
BC074953.2 | - | 1 | 195 | 30915500 | ||
X54667.1 | - | 1 | 222 |