Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSTF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001326.2 | NP_001317.1 | 18 | 1853 | ||
GenBank mRNA | AK290836.1 | - | 18 | 1853 | ||
AK294204.1 | - | 15 | 1446 | |||
AK297371.1 | - | 17 | 1735 | |||
BC010533.1 | - | 3 | 285 | |||
BC108319.1 | - | 18 | 1827 | 6197136 | ||
U15782.1 | - | 18 | 1818 |