Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKX2-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166175.1 | NP_001159647.1 | 2 | 1037 | ||
NM_001166176.1 | NP_001159648.1 | 2 | 1000 | |||
NM_004387.3 | NP_004378.1 | 2 | 745 | |||
GenBank mRNA | AB021133.1 | - | 2 | 516 | ||
AK290615.1 | - | 2 | 637 | |||
AK297844.1 | - | 2 | 894 | |||
AK307218.1 | - | 2 | 931 | |||
AK307249.1 | - | 2 | 930 | |||
AK309495.1 | - | 2 | 931 | |||
BC025711.1 | - | 2 | 623 | 5225103 | ||
KU178001.1 | - | 2 | 516 | |||
KU178002.1 | - | 2 | 808 | |||
U34962.1 | - | 2 | 692 |