Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280799.1 | NP_001267728.1 | 10 | 2757 | ||
NM_144698.4 | NP_653299.4 | 12 | 3027 | |||
GenBank mRNA | AK057853.1 | - | 3 | 917 | ||
AK091120.1 | - | 12 | 2992 | |||
AK294308.1 | - | 10 | 2736 | |||
BC101618.1 | - | 12 | 2974 | 8069124 | ||
BC101620.1 | - | 12 | 2974 | 8069126 |