Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181644.4 | NP_857595.3 | 9 | 1505 | ||
XM_011509229.2 | XP_011507531.1 | 8 | 1244 | |||
GenBank mRNA | AK057733.1 | - | 5 | 575 | ||
AK090831.1 | - | 3 | 232 | |||
AK091896.1 | - | 9 | 1446 | |||
AK127321.1 | - | 1 | 1141 | |||
AK293816.1 | - | 8 | 1434 | |||
AK295148.1 | - | 8 | 1092 | |||
AK304109.1 | - | 8 | 1310 | |||
AK304120.1 | - | 8 | 1171 | |||
AL713676.1 | - | 7 | 811 | |||
AY358107.1 | - | 7 | 892 | |||
BC036549.2 | - | 9 | 1455 | 5264261 |