Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181644.4 | NP_857595.3 | 3 | 673 | ||
GenBank mRNA | AK091896.1 | - | 3 | 614 | ||
AK293816.1 | - | 3 | 673 | |||
AK304109.1 | - | 2 | 478 | |||
AY358107.1 | - | 1 | 60 | |||
BC036549.2 | - | 3 | 623 | 5264261 |