Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024601.2 | NP_001019772.1 | 2 | 231 | ||
GenBank mRNA | AY640234.1 | - | 2 | 188 | ||
BC035742.1 | - | 2 | 231 | 5744157 |