Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C20orf196 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303477.1 | NP_001290406.1 | 3 | 315 | ||
NM_001303478.1 | NP_001290407.1 | 2 | 224 | |||
NM_001303479.1 | NP_001290408.1 | 2 | 224 | |||
NM_152504.3 | NP_689717.2 | 2 | 224 | |||
XM_006723549.3 | XP_006723612.1 | 2 | 155 | |||
XM_011529177.2 | XP_011527479.1 | 2 | 211 | |||
XM_011529178.2 | XP_011527480.1 | 2 | 262 | |||
XM_011529179.2 | XP_011527481.1 | 2 | 226 | |||
XM_011529180.2 | XP_011527482.1 | 2 | 221 | |||
XM_011529181.2 | XP_011527483.1 | 2 | 224 | |||
XM_011529182.2 | XP_011527484.1 | 2 | 224 | |||
XM_017027684.1 | XP_016883173.1 | 3 | 326 | |||
GenBank mRNA | AK057796.1 | - | 2 | 221 | ||
AK292708.1 | - | 2 | 210 | |||
AK310715.1 | - | 2 | 159 | |||
AV715251.1 | - | 2 | 209 | |||
BC035800.1 | - | 2 | 214 | 5734487 | ||
BU656896.1 | - | 3 | 268 | |||
CD694965.1 | - | 2 | 223 | |||
HY080554.1 | - | 2 | 217 |