Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318314.1 | NP_001305243.1 | 2 | 343 | ||
NM_001318315.1 | NP_001305244.1 | 2 | 343 | |||
NM_001318316.1 | NP_001305245.1 | 2 | 92 | |||
NM_172002.4 | NP_741999.3 | 2 | 343 | |||
XM_005261359.3 | XP_005261416.1 | 2 | 344 | |||
XM_017028619.1 | XP_016884108.1 | 2 | 309 | |||
XM_017028620.1 | XP_016884109.1 | 2 | 147 | |||
GenBank mRNA | AY191719.1 | - | 2 | 293 | ||
BC000004.1 | - | 2 | 352 | |||
BC065569.1 | - | 2 | 316 | 6171959 | ||
BQ054405.1 | - | 2 | 320 | |||
CR456462.1 | - | 2 | 311 | |||
CR979598.1 | - | 2 | 292 | |||
DW414316.1 | - | 2 | 108 | |||
HY057929.1 | - | 2 | 92 |