Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM109B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002034.2 | NP_001002034.2 | 3 | 447 | ||
XM_005261373.3 | XP_005261430.1 | 3 | 585 | |||
GenBank mRNA | BC029776.1 | - | 1 | 101 | ||
BC104175.1 | - | 2 | 343 | 40030677 | ||
BC104176.1 | - | 2 | 341 | 40030679 | ||
BX648402.1 | - | 3 | 447 | |||
CR456454.1 | - | 2 | 320 |