Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: KANSL1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307976.1 | NP_001294905.1 | 2 | 1127 | ||
NM_152519.3 | NP_689732.2 | 2 | 1127 | |||
XM_005246328.4 | XP_005246385.1 | 2 | 1409 | |||
XM_005246329.4 | XP_005246386.1 | 2 | 1115 | |||
XM_005246330.4 | XP_005246387.1 | 2 | 996 | |||
XM_005246332.4 | XP_005246389.1 | 2 | 1408 | |||
XM_005246334.4 | XP_005246391.1 | 2 | 1400 | |||
XM_006712320.3 | XP_006712383.1 | 2 | 1402 | |||
XM_011510706.2 | XP_011509008.1 | 2 | 1034 | |||
XM_011510707.2 | XP_011509009.1 | 2 | 1217 | |||
XM_011510709.2 | XP_011509011.1 | 2 | 1405 | |||
XM_017003430.1 | XP_016858919.1 | 2 | 1153 | |||
GenBank mRNA | AK056911.1 | - | 2 | 1014 | ||
AK124324.1 | - | 1 | 147 | |||
AK124963.1 | - | 2 | 1339 | |||
BC032837.1 | - | 2 | 1097 | 4829661 | ||
BC090854.1 | - | 1 | 465 | |||
BC126155.1 | - | 2 | 1033 | 8991871 | ||
BC143914.1 | - | 2 | 1033 | 9052432 |