Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033045.3 | NP_001028217.1 | 12 | 2251 | ||
NM_001267050.1 | NP_001253979.1 | 12 | 2148 | |||
NM_001267051.1 | NP_001253980.1 | 10 | 2018 | |||
NM_152529.6 | NP_689742.4 | 11 | 2102 | |||
XM_017003485.1 | XP_016858974.1 | 13 | 2293 | |||
XM_017003486.1 | XP_016858975.1 | 11 | 2154 | |||
XM_017003487.1 | XP_016858976.1 | 12 | 2155 | |||
GenBank mRNA | AK056381.1 | - | 9 | 1350 | ||
AK091200.1 | - | 11 | 2043 | |||
AK307896.1 | - | 11 | 2093 | |||
AK315125.1 | - | 11 | 2043 | |||
AL834154.1 | - | 12 | 2201 | |||
AY255528.1 | - | 2 | 259 | |||
BC028730.1 | - | 12 | 2197 | 4813561 | ||
BC035037.1 | - | 12 | 2159 | 5260361 | ||
BC036681.1 | - | 10 | 1964 | |||
BX537947.1 | - | 11 | 2095 |