Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGAT4D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277353.1 | NP_001264282.1 | 9 | 1045 | ||
XM_011531650.2 | XP_011529952.1 | 9 | 2497 | |||
XM_011531651.2 | XP_011529953.1 | 9 | 1218 | |||
XM_011531652.2 | XP_011529954.1 | 10 | 1143 | |||
XM_011531653.2 | XP_011529955.1 | 10 | 1075 | |||
XM_011531654.2 | XP_011529956.1 | 9 | 2440 | |||
XM_011531656.2 | XP_011529958.1 | 8 | 2359 | |||
XM_011531657.2 | XP_011529959.1 | 9 | 846 | |||
XM_011531658.2 | XP_011529960.1 | 7 | 2200 | |||
XM_011531659.1 | XP_011529961.1 | 8 | 2382 | |||
XM_011531661.1 | XP_011529963.1 | 7 | 2306 | |||
XM_011531662.1 | XP_011529964.1 | 6 | 2192 | |||
XM_011531663.2 | XP_011529965.1 | 7 | 2219 | |||
XM_011531664.2 | XP_011529966.1 | 6 | 780 | |||
XM_011531665.2 | XP_011529967.1 | 6 | 2060 | |||
GenBank mRNA | BC041818.1 | - | 6 | 3546 | ||
BI826002.1 | - | 3 | 396 | |||
BI828214.1 | - | 5 | 557 |