Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other JAKMIP1 SIRNA Assays ›
Gene Symbol:
JAKMIP1
Gene Name:
janus kinase and microtubule interacting protein 1
Gene Aliases:
Gababrbp, JAMIP1, MARLIN1
Chromosome Location:
Chr.4: 6024925 - 6200591 on Build GRCh38
UniGene ID:
Hs.479066
Species:
Human
siRNA ID
HSS175737
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: JAKMIP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001099433.1
NP_001092903.1
8, 9
1795
NM_001306133.1
NP_001293062.1
8, 9
1585
NM_001306134.1
NP_001293063.1
7, 8
1090
NM_144720.3
NP_653321.1
8, 9
1795
XM_011513400.2
XP_011511702.1
7, 8
1264
XM_017007788.1
XP_016863277.1
8, 9
1759
XM_017007789.1
XP_016863278.1
8, 9
1583
XM_017007790.1
XP_016863279.1
8, 9
1735
XM_017007791.1
XP_016863280.1
8, 9
1759
XM_017007792.1
XP_016863281.1
8, 9
1759
XM_017007793.1
XP_016863282.1
8, 9
1759
XM_017007794.1
XP_016863283.1
8, 9
1759
XM_017007795.1
XP_016863284.1
8, 9
1759
XM_017007796.1
XP_016863285.1
8, 9
1759
XM_017007797.1
XP_016863286.1
7, 8
1264
XM_017007798.1
XP_016863287.1
7, 8
1902
XM_017007799.1
XP_016863288.1
7, 8
1264
GenBank mRNA
AK056126.1
-
8, 9
1759
AK098536.1
-
6, 7
869
AK124718.1
-
8, 9
1583
AK294356.1
-
9, 10
1412
AK295339.1
-
7, 8
1090
AK307983.1
-
7, 8
1010
AM412307.1
-
8, 9
1759
AM412309.1
-
8, 9
1759
AY382340.1
-
7, 8
1309
BC047075.1
-
8, 9
1515
5314069
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein transport
cognition
RNA binding
protein binding
microtubule binding
kinesin binding
kinase binding
GABA receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbcvc:80/10.239.164.135:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline