Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMOT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113490.1 | NP_001106962.1 | 8 | 2133 | ||
NM_133265.2 | NP_573572.1 | 9 | 1702 | |||
XM_005262087.1 | XP_005262144.1 | 11 | 2878 | |||
XM_005262090.1 | XP_005262147.1 | 8 | 1375 | |||
XM_011530875.2 | XP_011529177.1 | 12 | 2990 | |||
XM_017029289.1 | XP_016884778.1 | 10 | 1906 | |||
GenBank mRNA | AB028994.3 | - | 7 | 906 | ||
AF286598.1 | - | 9 | 1702 | |||
AY987378.1 | - | 8 | 2133 | |||
BC130294.1 | - | 7 | 1123 | 40146325 |