Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000026.3 | NP_000017.1 | 2 | 381 | ||
NM_001123378.2 | NP_001116850.1 | 2 | 381 | |||
NM_001317923.1 | NP_001304852.1 | 2 | 326 | |||
NR_134256.1 | 2 | 381 | ||||
XM_011529977.2 | XP_011528279.1 | 2 | 381 | |||
XM_011529979.1 | XP_011528281.1 | 2 | 381 | |||
XM_011529980.2 | XP_011528282.1 | 2 | 381 | |||
XM_017028636.1 | XP_016884125.1 | 2 | 363 | |||
XM_017028637.1 | XP_016884126.1 | 2 | 346 | |||
XM_017028638.1 | XP_016884127.1 | 2 | 363 | |||
XM_017028639.1 | XP_016884128.1 | 2 | 363 | |||
GenBank mRNA | AF067853.1 | - | 2 | 377 | ||
AF067854.1 | - | 2 | 377 | |||
AK225042.1 | - | 2 | 349 | |||
AK291091.1 | - | 2 | 352 | |||
AK293722.1 | - | 2 | 363 | |||
AK298187.1 | - | 2 | 353 | |||
BC000253.1 | - | 2 | 351 | 3352079 | ||
BC018881.2 | - | 2 | 351 | |||
BP298407.1 | - | 2 | 381 | |||
CR456368.1 | - | 2 | 355 | |||
KJ534763.1 | - | 2 | 322 | |||
KJ535007.1 | - | 2 | 322 | |||
KJ535020.1 | - | 2 | 322 | |||
KJ535056.1 | - | 2 | 267 | |||
S60710.1 | - | 2 | 324 | |||
X65867.1 | - | 2 | 324 |