Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DGKA SIRNA Assays ›
Gene Symbol:
DGKA
Gene Name:
diacylglycerol kinase alpha
Gene Aliases:
DAGK, DAGK1, DGK-alpha
Chromosome Location:
Chr.12: 55931076 - 55954023 on Build GRCh38
UniGene ID:
Hs.524488
Species:
Human
siRNA ID
HSS175915
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DGKA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001345.4
NP_001336.2
12, 13
1288
NM_201444.2
NP_958852.1
12, 13
1333
NM_201445.1
NP_958853.1
13, 14
1301
NM_201554.1
NP_963848.1
12, 13
1201
XM_005268688.2
XP_005268745.1
12, 13
1230
XM_005268689.2
XP_005268746.1
12, 13
1165
XM_005268690.2
XP_005268747.1
10, 11
985
XM_011537991.2
XP_011536293.1
13, 14
1398
XM_011537993.2
XP_011536295.1
12, 13
1149
XM_011537995.2
XP_011536297.1
12, 13
1160
XM_017018900.1
XP_016874389.1
11, 12
1584
XM_017018901.1
XP_016874390.1
11, 12
1728
XM_017018902.1
XP_016874391.1
11, 12
1550
XM_017018903.1
XP_016874392.1
12, 13
1752
XM_017018904.1
XP_016874393.1
12, 13
1620
XM_017018905.1
XP_016874394.1
13, 14
2029
XM_017018906.1
XP_016874395.1
11, 12
1661
XM_017018907.1
XP_016874396.1
11, 12
1484
XM_017018908.1
XP_016874397.1
12, 13
1368
XM_017018909.1
XP_016874398.1
11, 12
1119
GenBank mRNA
AF064771.1
-
12, 13
1246
AK122973.1
-
10, 11
2639
AK127093.1
-
4, 5
1976
AK226134.1
-
7, 8
2850
AK303320.1
-
11, 12
1416
AK307685.1
-
11, 12
1045
AY930112.1
-
11, 12
1139
BC023523.2
-
13, 14
1224
4304187
BC031870.2
-
12, 13
1228
4819985
X62535.1
-
11, 12
1127
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
Gene Ontology Categories:
Function(s)
Process(es)
phosphatidic acid biosynthetic process
protein kinase C-activating G-protein coupled receptor signaling pathway
phosphorylation
platelet activation
intracellular signal transduction
diacylglycerol kinase activity
calcium ion binding
ATP binding
phospholipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-sshrn:80/10.239.165.44:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0