Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153228.2 | NP_694960.2 | 17 | 2276 | ||
XM_006721728.3 | XP_006721791.1 | 16 | 2499 | |||
XM_011524428.2 | XP_011522730.1 | 17 | 2276 | |||
XM_011524429.2 | XP_011522731.1 | 17 | 2433 | |||
XM_011524430.2 | XP_011522732.1 | 18 | 2399 | |||
XM_011524431.2 | XP_011522733.1 | 17 | 2331 | |||
XM_011524433.2 | XP_011522735.1 | 11 | 1689 | |||
XM_011524434.2 | XP_011522736.1 | 8 | 1136 | |||
XM_017024263.1 | XP_016879752.1 | 19 | 2493 | |||
XM_017024264.1 | XP_016879753.1 | 19 | 3478 | |||
XM_017024265.1 | XP_016879754.1 | 19 | 2687 | |||
XM_017024266.1 | XP_016879755.1 | 17 | 2448 | |||
XM_017024267.1 | XP_016879756.1 | 16 | 2300 | |||
XM_017024268.1 | XP_016879757.1 | 18 | 3384 | |||
XM_017024269.1 | XP_016879758.1 | 16 | 2573 | |||
XM_017024270.1 | XP_016879759.1 | 19 | 2491 | |||
GenBank mRNA | AK095654.1 | - | 17 | 2276 |