Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF781 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152605.3 | NP_689818.2 | 4 | 789 | ||
GenBank mRNA | AK097019.1 | - | 4 | 725 | ||
BC108687.1 | - | 4 | 709 | 6148649 |