Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TTLL9 SIRNA Assays ›
Gene Symbol:
TTLL9
Gene Name:
tubulin tyrosine ligase like 9
Gene Aliases:
C20orf125
Chromosome Location:
Chr.20: 31870702 - 31944959 on Build GRCh38
UniGene ID:
Hs.712915
Species:
Human
siRNA ID
HSS176023
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TTLL9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001008409.2
NP_001008409.1
5
513
NR_134519.1
6
599
XM_005260309.2
XP_005260366.1
4
277
XM_006723723.2
XP_006723786.1
3
214
XM_006723724.2
XP_006723787.1
4
386
XM_011528617.1
XP_011526919.1
5
349
XM_011528618.1
XP_011526920.1
5
513
XM_011528619.1
XP_011526921.1
6
411
XM_011528620.2
XP_011526922.1
6
408
XM_011528621.1
XP_011526923.1
6
426
XM_011528622.1
XP_011526924.1
6
401
XM_011528623.2
XP_011526925.1
4
282
XM_011528624.1
XP_011526926.1
5
349
XM_011528625.1
XP_011526927.1
5
349
XM_011528626.1
XP_011526928.1
5
347
XM_011528627.2
XP_011526929.1
5
344
XM_011528628.1
XP_011526930.1
4
272
XM_011528629.1
XP_011526931.1
4
280
XM_011528630.1
XP_011526932.1
3
214
XM_011528631.1
XP_011526933.1
5
349
XM_011528632.1
XP_011526934.1
5
349
XM_011528633.1
XP_011526935.1
5
349
XM_011528634.1
XP_011526936.1
5
349
XM_011528635.1
XP_011526937.1
5
349
XM_011528637.2
XP_011526939.1
5
349
XM_011528638.1
XP_011526940.1
5
349
XM_011528639.1
XP_011526941.1
5
349
XM_011528640.1
XP_011526942.1
5
349
XM_011528641.1
XP_011526943.1
5
349
XM_011528643.2
XP_011526945.1
5
349
XM_011528644.1
XP_011526946.1
5
349
XM_011528646.1
XP_011526948.1
6
538
XM_017027694.1
XP_016883183.1
3
214
XM_017027695.1
XP_016883184.1
5
349
XM_017027696.1
XP_016883185.1
5
349
XM_017027697.1
XP_016883186.1
5
349
XM_017027698.1
XP_016883187.1
5
349
XM_017027699.1
XP_016883188.1
4
532
XM_017027700.1
XP_016883189.1
5
349
XM_017027701.1
XP_016883190.1
5
349
XM_017027702.1
XP_016883191.1
5
349
XM_017027703.1
XP_016883192.1
5
349
GenBank mRNA
AK093491.1
-
5
514
AK302360.1
-
3
216
AK302806.1
-
4
260
BC068593.1
-
6
590
BC104024.1
-
6
538
40025306
BC104025.1
-
4
386
40025308
Back To Top
More Information
Panther Classification:
Molecular Function -
microtubule or microtubule-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
cellular protein modification process
ATP binding
ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-9xb8z:80/10.239.165.207:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0