Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PTCRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243168.1 | NP_001230097.1 | 4 | 904 | ||
NM_001243169.1 | NP_001230098.1 | 5 | 784 | |||
NM_001243170.1 | NP_001230099.1 | 3 | 538 | |||
NM_138296.2 | NP_612153.2 | 4 | 859 | |||
GenBank mRNA | AF101436.1 | - | 3 | 457 | ||
AF165312.1 | - | 5 | 703 | |||
AK303523.1 | - | 4 | 334 | |||
AK312223.1 | - | 4 | 792 | |||
AW014894.1 | - | 1 | 235 | |||
BC069336.1 | - | 4 | 827 | 7262230 | ||
BC100771.1 | - | 4 | 801 | 40000277 | ||
BC100772.1 | - | 4 | 801 | 40000280 | ||
BC100773.2 | - | 4 | 801 | 40000282 | ||
BC100774.2 | - | 4 | 801 | 40000286 | ||
BC153829.1 | - | 4 | 801 | 40000274 | ||
U36759.1 | - | 4 | 859 | |||
U38996.1 | - | 4 | 744 |