Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNASE1L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004944.3 | NP_004935.1 | 3, 4 | 408 | ||
GenBank mRNA | AF047354.1 | - | 3, 4 | 381 | ||
AK131323.1 | - | 6, 7 | 903 | |||
AK313303.1 | - | 3, 4 | 374 | |||
BC015831.1 | - | 3, 4 | 378 | 4723725 | ||
BI521582.1 | - | 3, 4 | 618 | |||
U56814.1 | - | 3, 4 | 335 |