Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201478.1 | NP_001188407.1 | 11 | 1326 | ||
NM_006893.2 | NP_008824.2 | 13 | 1698 | |||
XM_005277509.3 | XP_005277566.1 | 12 | 1463 | |||
XM_006711196.3 | XP_006711259.1 | 13 | 1569 | |||
XM_011509257.2 | XP_011507559.1 | 13 | 1613 | |||
XM_017000530.1 | XP_016856019.1 | 10 | 1133 | |||
XM_017000531.1 | XP_016856020.1 | 10 | 1091 | |||
XM_017000532.1 | XP_016856021.1 | 12 | 1419 | |||
GenBank mRNA | AB209364.1 | - | 4 | 3526 | ||
AF220417.2 | - | 13 | 1606 | |||
AF262403.1 | - | 11 | 1234 | |||
AK001852.1 | - | 13 | 1611 | |||
AK225259.1 | - | 13 | 1592 | |||
AL162001.1 | - | 8 | 904 | |||
BC001585.1 | - | 13 | 1596 | 3463026 | ||
BC039134.1 | - | 13 | 1549 | 4339127 | ||
BC058905.1 | - | 13 | 1563 | 6067337 |