Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153486.3 | NP_705690.2 | 4 | 394 | ||
NM_194436.2 | NP_919417.1 | 4 | 394 | |||
GenBank mRNA | AY092767.1 | - | 4 | 399 | ||
BC040279.1 | - | 4 | 359 | 5162826 | ||
BC047902.1 | - | 4 | 389 | 5762691 | ||
BM544922.1 | - | 4 | 400 |