Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152670.2 | NP_689883.1 | 3 | 183 | ||
XM_005264188.3 | XP_005264245.3 | 3 | 158 | |||
XM_011532691.1 | XP_011530993.1 | 4 | 277 | |||
XM_011532692.1 | XP_011530994.1 | 3 | 211 | |||
GenBank mRNA | AK058098.1 | - | 3 | 183 | ||
BC029522.1 | - | 3 | 150 | 5297675 |