Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ELN SIRNA Assays ›
Gene Symbol:
ELN
Gene Name:
elastin
Gene Aliases:
SVAS, WBS, WS
Chromosome Location:
Chr.7: 74027772 - 74069907 on Build GRCh38
UniGene ID:
Hs.647061
Species:
Human
siRNA ID
HSS176467
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ELN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000501.3
NP_000492.2
29
2365
NM_001081752.2
NP_001075221.1
27
2278
NM_001081753.2
NP_001075222.1
28
2323
NM_001081754.2
NP_001075223.1
29
2380
NM_001081755.2
NP_001075224.1
28
2308
NM_001278912.1
NP_001265841.1
29
2365
NM_001278913.1
NP_001265842.1
25
2122
NM_001278914.1
NP_001265843.1
27
2293
NM_001278915.1
NP_001265844.1
29
2383
NM_001278916.1
NP_001265845.1
26
2221
NM_001278917.1
NP_001265846.1
28
2335
NM_001278918.1
NP_001265847.1
25
2098
NM_001278939.1
NP_001265868.1
30
2551
XM_005250187.1
XP_005250244.1
28
2006
XM_005250188.1
XP_005250245.1
28
1998
XM_011515868.1
XP_011514170.1
29
2066
XM_011515869.1
XP_011514171.1
28
1995
XM_011515870.1
XP_011514172.1
28
2361
XM_011515871.1
XP_011514173.1
28
2024
XM_011515872.1
XP_011514174.1
28
2012
XM_011515873.1
XP_011514175.1
28
2009
XM_011515874.1
XP_011514176.1
27
2331
XM_011515875.1
XP_011514177.1
27
1987
XM_011515876.1
XP_011514178.1
29
2066
XM_011515877.1
XP_011514179.1
27
1955
XM_017011813.1
XP_016867302.1
27
1909
XM_017011814.1
XP_016867303.1
27
1967
GenBank mRNA
AB208942.1
-
28
1961
AK075554.1
-
25
1805
AK092232.1
-
25
1713
AK094138.1
-
29
1798
AK095990.1
-
28
1943
AK122731.1
-
22
1571
AK125511.1
-
27
1951
AK130894.1
-
15
914
AK225659.1
-
28
1922
AK304845.1
-
29
2365
BC035570.1
-
29
1970
BC050379.1
-
23
1293
BC065566.1
-
26
1852
6153564
BX537939.1
-
29
2000
BX538199.1
-
27
1910
M24782.1
-
20
1477
M36860.1
-
29
2033
X52896.1
-
29
2032
Back To Top
More Information
Panther Classification:
Molecular Function -
structural protein
Gene Ontology Categories:
Function(s)
Process(es)
respiratory gaseous exchange
blood circulation
cell proliferation
organ morphogenesis
extracellular matrix disassembly
extracellular matrix organization
extracellular matrix structural constituent
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0