Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM184A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097620.1 | NP_001091089.1 | 5 | 845 | ||
XM_006715653.1 | XP_006715716.1 | 5 | 845 | |||
XM_006715655.2 | XP_006715718.1 | 5 | 672 | |||
XM_011515164.1 | XP_011513466.1 | 5 | 845 | |||
XM_011515165.2 | XP_011513467.1 | 5 | 845 | |||
XM_011515166.2 | XP_011513468.1 | 5 | 845 | |||
XM_017011815.1 | XP_016867304.1 | 5 | 672 | |||
GenBank mRNA | AK096790.1 | - | 3 | 241 | ||
AK172850.1 | - | 5 | 621 | |||
AK313249.1 | - | 5 | 629 | |||
BC026694.1 | - | 5 | 620 | 3850351 |