Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293212.1 | NP_001280141.1 | 4 | 954 | ||
NM_001293214.1 | NP_001280143.1 | 3 | 738 | |||
NM_001293215.1 | NP_001280144.1 | 4 | 857 | |||
NM_001293216.1 | NP_001280145.1 | 4 | 756 | |||
NM_178014.3 | NP_821133.1 | 4 | 870 | |||
GenBank mRNA | AB062393.1 | - | 4 | 696 | ||
AF070561.1 | - | 4 | 691 | |||
AF070593.1 | - | 4 | 691 | |||
AF070600.1 | - | 4 | 708 | |||
AF141349.1 | - | 4 | 708 | |||
AK298889.1 | - | 4 | 760 | |||
AK302314.1 | - | 4 | 823 | |||
AK303227.1 | - | 3 | 457 | |||
AK309684.1 | - | 2 | 312 | |||
AK316265.1 | - | 4 | 627 | |||
AK316316.1 | - | 4 | 752 | |||
BC001896.1 | - | 1 | 252 | |||
BC001938.1 | - | 4 | 695 | 2820585 | ||
BC002347.2 | - | 4 | 700 | 2964461 | ||
BC005838.2 | - | 4 | 695 | 2820585 | ||
BC007605.1 | - | 4 | 700 | 3350604 | ||
BC013374.2 | - | 4 | 691 | 3946253 | ||
BC015889.1 | - | 1 | 32 | |||
BC019924.1 | - | 4 | 688 | 4998571 | ||
BC020946.1 | - | 4 | 729 | 4795692 | ||
BC021909.1 | - | 4 | 690 | 3509417 | ||
BC029323.1 | - | 1 | 53 | |||
BC070326.1 | - | 4 | 706 | 6698679 | ||
BC103746.1 | - | 4 | 713 | 6503977 | ||
DQ457058.1 | - | 1 | 126 | |||
V00599.1 | - | 3 | 543 |