Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCDC171 SIRNA Assays ›
Gene Symbol:
CCDC171
Gene Name:
coiled-coil domain containing 171
Gene Aliases:
C9orf93, bA536D16.1, bA778P13.1
Chromosome Location:
Chr.9: 15552874 - 16062431 on Build GRCh38
UniGene ID:
Hs.17267
Species:
Human
siRNA ID
HSS176558
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCDC171
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_173550.2
NP_775821.2
14
1883
XM_005251397.2
XP_005251454.1
14
1790
XM_005251398.3
XP_005251455.1
14
2029
XM_005251399.4
XP_005251456.1
15
2089
XM_011517788.2
XP_011516090.1
14
2019
XM_011517791.2
XP_011516093.1
13
1897
XM_011517796.2
XP_011516098.1
12
1436
XM_017014431.1
XP_016869920.1
14
2029
XM_017014432.1
XP_016869921.1
14
2029
XM_017014433.1
XP_016869922.1
14
2029
XM_017014434.1
XP_016869923.1
14
2029
XM_017014435.1
XP_016869924.1
14
2029
XM_017014436.1
XP_016869925.1
14
2029
XM_017014437.1
XP_016869926.1
14
2029
XM_017014438.1
XP_016869927.1
14
2028
XM_017014439.1
XP_016869928.1
15
2191
XM_017014440.1
XP_016869929.1
12
1450
XM_017014441.1
XP_016869930.1
14
2028
XM_017014442.1
XP_016869931.1
14
2028
XM_017014443.1
XP_016869932.1
13
1511
XM_017014444.1
XP_016869933.1
3
230
GenBank mRNA
AK128581.1
-
2
146
AK308284.1
-
14
1790
AY422473.1
-
14
1883
BC064849.1
-
1
89
BC144214.1
-
13
1684
9052734
BC150600.1
-
13
1684
9056970
CR936775.1
-
15
2161
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0