Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DMTN SIRNA Assays ›
Gene Symbol:
DMTN
Gene Name:
dematin actin binding protein
Gene Aliases:
DMT, EPB49
Chromosome Location:
Chr.8: 22048955 - 22082527 on Build GRCh38
UniGene ID:
Hs.106124
Species:
Human
siRNA ID
HSS176582
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DMTN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001114135.4
NP_001107607.1
8
759
NM_001114136.2
NP_001107608.1
8
715
NM_001114137.3
NP_001107609.1
8
889
NM_001114138.2
NP_001107610.1
8
876
NM_001114139.3
NP_001107611.1
7
773
NM_001302816.2
NP_001289745.1
8
1009
NM_001302817.2
NP_001289746.1
6
728
NM_001323378.1
NP_001310307.1
8
848
NM_001323380.1
NP_001310309.1
8
755
NM_001323381.1
NP_001310310.1
7
864
NM_001323382.1
NP_001310311.1
8
794
NM_001323383.1
NP_001310312.1
8
715
NM_001323384.1
NP_001310313.1
8
848
NM_001323385.1
NP_001310314.1
7
864
NM_001323387.1
NP_001310316.1
7
813
NM_001323388.1
NP_001310317.1
6
595
NM_001323389.1
NP_001310318.1
7
803
NM_001323390.1
NP_001310319.1
7
670
NM_001323391.1
NP_001310320.1
7
773
NM_001323392.1
NP_001310321.1
7
934
NM_001323393.1
NP_001310322.1
7
640
NM_001323394.1
NP_001310323.1
7
801
NM_001323395.1
NP_001310324.1
7
680
NM_001323396.1
NP_001310325.1
7
684
NM_001323397.1
NP_001310326.1
7
852
NM_001323398.1
NP_001310327.1
6
728
NM_001323399.1
NP_001310328.1
6
595
NM_001323400.1
NP_001310329.1
5
744
NM_001323401.1
NP_001310330.1
6
768
NM_001978.4
NP_001969.2
8
1009
XM_005273432.1
XP_005273489.1
8
741
XM_005273433.1
XP_005273490.1
8
673
XM_005273442.1
XP_005273499.1
7
525
XM_011544435.1
XP_011542737.1
7
934
XM_017013189.1
XP_016868678.1
8
682
XM_017013190.1
XP_016868679.1
7
934
XM_017013191.1
XP_016868680.1
7
688
XM_017013192.1
XP_016868681.1
7
569
XM_017013193.1
XP_016868682.1
6
701
XM_017013194.1
XP_016868683.1
6
568
XM_017013195.1
XP_016868684.1
7
934
XM_017013196.1
XP_016868685.1
7
537
XM_017013197.1
XP_016868686.1
7
768
XM_017013198.1
XP_016868687.1
7
610
GenBank mRNA
AB209718.1
-
4
235
AK055842.1
-
8
673
AK091581.1
-
7
688
AK289650.1
-
8
641
AK295452.1
-
6
614
AK308807.1
-
6
908
BC006318.2
-
8
758
4125804
BC017445.1
-
8
758
3162732
BC052805.1
-
8
799
6269658
BM547218.1
-
7
831
BT007396.1
-
7
486
L19713.1
-
8
936
U28389.1
-
8
936
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
protein complex assembly
cytoskeleton organization
regulation of cell shape
regulation of lamellipodium assembly
positive regulation of fibroblast migration
negative regulation of peptidyl-threonine phosphorylation
negative regulation of cell-substrate adhesion
actin cytoskeleton organization
positive regulation of blood coagulation
regulation of actin cytoskeleton organization
negative regulation of peptidyl-serine phosphorylation
calcium-mediated signaling using intracellular calcium source
calcium-mediated signaling using extracellular calcium source
erythrocyte development
negative regulation of peptidyl-tyrosine phosphorylation
actin filament bundle assembly
regulation of filopodium assembly
actin filament capping
negative regulation of focal adhesion assembly
transmembrane transport
protein secretion by platelet
cellular response to calcium ion
cellular response to cAMP
positive regulation of wound healing
negative regulation of protein targeting to membrane
actin filament reorganization
negative regulation of substrate adhesion-dependent cell spreading
positive regulation of substrate adhesion-dependent cell spreading
positive regulation of platelet aggregation
positive regulation of integrin-mediated signaling pathway
actin binding
receptor binding
protein binding
spectrin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-gnqd7:80/10.239.161.169:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline