Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EWSR1 SIRNA Assays ›
Gene Symbol:
EWSR1
Gene Name:
EWS RNA binding protein 1
Gene Aliases:
EWS, EWS-FLI1, bK984G1.4
Chromosome Location:
Chr.22: 29268009 - 29300525 on Build GRCh38
UniGene ID:
Hs.374477
Species:
Human
siRNA ID
HSS176640
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EWSR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001163285.1
NP_001156757.1
11
1403
NM_001163286.1
NP_001156758.1
10
1238
NM_005243.3
NP_005234.1
11
1406
NM_013986.3
NP_053733.2
12
1421
XM_005261389.3
XP_005261446.1
11
1161
XM_005261390.4
XP_005261447.1
10
993
XM_011529995.2
XP_011528297.1
11
1136
XM_011529996.2
XP_011528298.1
11
1133
XM_011529997.1
XP_011528299.1
11
1136
XM_011529998.1
XP_011528300.1
11
1133
XM_011529999.2
XP_011528301.1
11
1136
XM_011530000.1
XP_011528302.1
11
1136
XM_011530001.2
XP_011528303.1
10
968
XM_011530002.2
XP_011528304.1
9
917
XM_017028644.1
XP_016884133.1
11
1158
XM_017028645.1
XP_016884134.1
11
1161
XM_017028646.1
XP_016884135.1
11
1133
XM_017028647.1
XP_016884136.1
11
1158
XM_017028648.1
XP_016884137.1
11
1158
XM_017028649.1
XP_016884138.1
11
1136
XM_017028650.1
XP_016884139.1
11
1136
XM_017028651.1
XP_016884140.1
11
1161
XM_017028652.1
XP_016884141.1
11
1133
XM_017028653.1
XP_016884142.1
11
1158
XM_017028654.1
XP_016884143.1
10
965
XM_017028655.1
XP_016884144.1
10
990
XM_017028656.1
XP_016884145.1
11
1161
XM_017028657.1
XP_016884146.1
11
4887
XM_017028658.1
XP_016884147.1
10
965
XM_017028659.1
XP_016884148.1
10
990
XM_017028660.1
XP_016884149.1
11
1158
XM_017028661.1
XP_016884150.1
11
4884
XM_017028662.1
XP_016884151.1
10
990
XM_017028663.1
XP_016884152.1
10
990
XM_017028664.1
XP_016884153.1
10
1078
XM_017028665.1
XP_016884154.1
10
1078
XM_017028666.1
XP_016884155.1
10
4849
GenBank mRNA
AK026270.1
-
2
1374
AK056309.1
-
12
1114
AK056681.1
-
10
921
AK127624.1
-
7
959
AK225096.1
-
11
1085
AK225102.1
-
11
1097
AL833489.1
-
3
3030
BC004817.1
-
11
1097
3449145
BC011048.1
-
11
1102
4340116
BC072442.1
-
10
1081
6047805
BE741903.1
-
2
103
BX648769.1
-
6
6028
CR456490.1
-
11
1108
JX977847.1
-
11
1298
X66899.1
-
11
1121
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
nucleotide binding
RNA binding
protein binding
calmodulin binding
zinc ion binding
identical protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0