Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EWSR1 SIRNA Assays ›
Gene Symbol:
EWSR1
Gene Name:
EWS RNA binding protein 1
Gene Aliases:
EWS, EWS-FLI1, bK984G1.4
Chromosome Location:
Chr.22: 29268009 - 29300525 on Build GRCh38
UniGene ID:
Hs.374477
Species:
Human
siRNA ID
HSS176641
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EWSR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001163285.1
NP_001156757.1
4
471
NM_001163286.1
NP_001156758.1
4
471
NM_001163287.1
NP_001156759.1
4
471
NM_005243.3
NP_005234.1
4
471
NM_013986.3
NP_053733.2
4
471
XM_005261389.3
XP_005261446.1
4
226
XM_005261390.4
XP_005261447.1
4
226
XM_011529995.2
XP_011528297.1
4
201
XM_011529996.2
XP_011528298.1
4
201
XM_011529997.1
XP_011528299.1
4
201
XM_011529998.1
XP_011528300.1
4
201
XM_011529999.2
XP_011528301.1
4
201
XM_011530000.1
XP_011528302.1
4
201
XM_011530001.2
XP_011528303.1
4
201
XM_011530002.2
XP_011528304.1
4
201
XM_017028644.1
XP_016884133.1
4
226
XM_017028645.1
XP_016884134.1
4
226
XM_017028646.1
XP_016884135.1
4
201
XM_017028647.1
XP_016884136.1
4
226
XM_017028648.1
XP_016884137.1
4
226
XM_017028649.1
XP_016884138.1
4
201
XM_017028650.1
XP_016884139.1
4
201
XM_017028651.1
XP_016884140.1
4
226
XM_017028652.1
XP_016884141.1
4
201
XM_017028653.1
XP_016884142.1
4
226
XM_017028654.1
XP_016884143.1
4
201
XM_017028655.1
XP_016884144.1
4
226
XM_017028656.1
XP_016884145.1
4
226
XM_017028657.1
XP_016884146.1
4
187
XM_017028658.1
XP_016884147.1
4
201
XM_017028659.1
XP_016884148.1
4
226
XM_017028660.1
XP_016884149.1
4
226
XM_017028661.1
XP_016884150.1
4
187
XM_017028662.1
XP_016884151.1
4
226
XM_017028663.1
XP_016884152.1
4
226
XM_017028664.1
XP_016884153.1
4
181
XM_017028665.1
XP_016884154.1
4
181
XM_017028666.1
XP_016884155.1
4
187
GenBank mRNA
AK056309.1
-
4
164
AK056681.1
-
4
154
AK127624.1
-
180
AK225096.1
-
4
153
AK225102.1
-
4
165
BC000527.2
-
4
183
2961279
BC004817.1
-
4
162
3449145
BC011048.1
-
4
170
4340116
BC072442.1
-
3
149
6047805
CR456490.1
-
4
176
CX786322.1
-
4
201
DA013931.1
-
4
465
DB051511.1
-
4
452
JX977847.1
-
4
471
X66899.1
-
4
186
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
nucleotide binding
RNA binding
protein binding
calmodulin binding
zinc ion binding
identical protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0