Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000477.5 | NP_000468.1 | 7 | 893 | ||
GenBank mRNA | A06977.1 | - | 7 | 895 | ||
AF190168.1 | - | 7 | 820 | |||
AF542069.1 | - | 7 | 853 | |||
AK292755.1 | - | 7 | 858 | |||
AK298437.1 | - | 7 | 886 | |||
AK298461.1 | - | 7 | 603 | |||
AK308044.1 | - | 7 | 736 | |||
AK314794.1 | - | 7 | 861 | |||
AY544124.1 | - | 3 | 251 | |||
AY550967.1 | - | 7 | 827 | |||
AY728024.1 | - | 7 | 826 | |||
AY960291.1 | - | 7 | 852 | |||
BC034023.1 | - | 7 | 861 | 4734617 | ||
BC036003.1 | - | 7 | 861 | 4724105 | ||
BC041789.1 | - | 3 | 285 | 4766983 | ||
CR749331.1 | - | 7 | 860 | |||
DQ986150.1 | - | 7 | 831 | |||
HQ537426.1 | - | 6 | 748 | |||
V00494.1 | - | 7 | 859 | |||
V00495.1 | - | 7 | 895 |