Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152721.5 | NP_689934.2 | 6 | 854 | ||
XM_011525875.2 | XP_011524177.1 | 6 | 875 | |||
XM_017025610.1 | XP_016881099.1 | 4 | 2915 | |||
XM_017025611.1 | XP_016881100.1 | 4 | 1429 | |||
GenBank mRNA | AK057795.1 | - | 5 | 563 | ||
AY599248.1 | - | 6 | 852 | |||
BC008583.1 | - | 4 | 436 | |||
BC019045.2 | - | 4 | 479 | |||
BC096744.1 | - | 6 | 847 | 5260167 |