Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALKBH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139178.3 | NP_631917.1 | 6, 7 | 810 | ||
GenBank mRNA | AB042029.1 | - | 6, 7 | 773 | ||
BC015155.1 | - | 1, 2 | 96 | |||
BC103812.1 | - | 6, 7 | 548 | 40000372 | ||
BC103813.1 | - | 6, 7 | 548 | 40000375 | ||
BC103814.2 | - | 6, 7 | 548 | 40000376 |